siRNA Details
Gene Symbol: BRCA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007294.3 | NP_009225.1 | 4, 5 | 452 | ||
NM_007297.3 | NP_009228.2 | 3, 4 | 360 | |||
NM_007298.3 | NP_009229.2 | 3, 4 | 239 | |||
NM_007299.3 | NP_009230.2 | 4, 5 | 414 | |||
NM_007300.3 | NP_009231.2 | 4, 5 | 452 | |||
GenBank mRNA | AK308084.1 | - | 4, 5 | 381 | ||
AK316200.1 | - | 3, 4 | 311 | |||
AY354539.2 | - | 3, 4 | 284 | |||
AY751490.1 | - | 3, 4 | 220 | |||
BC030969.1 | - | 4, 5 | 339 | |||
BC072418.1 | - | 4, 5 | 414 | |||
BC085615.1 | - | 4, 5 | 322 | |||
BC106745.1 | - | 4, 5 | 269 | |||
BC114511.1 | - | 5, 6 | 385 | |||
BC115037.1 | - | 4, 5 | 357 | 7961446 | ||
JN686490.1 | - | 3, 4 | 220 | |||
KP272105.1 | - | 140 | ||||
U14680.1 | - | 4, 5 | 339 | |||
U64805.1 | - | 3, 4 | 220 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
