siRNA Details
Gene Symbol: CCNA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111045.1 | NP_001104515.1 | 3 | 669 | ||
NM_001111046.1 | NP_001104516.1 | 3 | 551 | |||
NM_001111047.1 | NP_001104517.1 | 3 | 548 | |||
NM_003914.3 | NP_003905.1 | 3 | 672 | |||
XM_011535294.2 | XP_011533596.1 | 3 | 1503 | |||
XM_011535295.2 | XP_011533597.1 | 3 | 1500 | |||
XM_011535296.2 | XP_011533598.1 | 3 | 328 | |||
GenBank mRNA | AK225146.1 | - | 3 | 449 | ||
AK301897.1 | - | 3 | 444 | |||
AK314261.1 | - | 3 | 672 | |||
AK316392.1 | - | 3 | 471 | |||
BC035479.1 | - | 3 | 418 | |||
BC036346.1 | - | 3 | 418 | 5172478 | ||
BG723613.1 | - | 3 | 554 | |||
BT019577.1 | - | 3 | 313 | |||
CD171793.1 | - | 3 | 471 | |||
DC395631.1 | - | 3 | 431 | |||
U66838.1 | - | 3 | 451 | |||
U97680.1 | - | 2 | 161 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
