siRNA Details
Gene Symbol: CCNB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031966.3 | NP_114172.1 | 2, 3 | 453 | ||
GenBank mRNA | AK289431.1 | - | 2, 3 | 342 | ||
AM851053.1 | - | 2, 3 | 200 | |||
BC006510.2 | - | 2, 3 | 295 | 2963100 | ||
BI253686.1 | - | 2, 3 | 453 | |||
BT020128.1 | - | 2, 3 | 200 | |||
M25753.1 | - | 2, 3 | 289 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
