siRNA Details
Gene Symbol: CCNE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001238.3 | NP_001229.1 | 6 | 560 | ||
NM_001322259.1 | NP_001309188.1 | 6 | 560 | |||
NM_001322261.1 | NP_001309190.1 | 6 | 560 | |||
NM_001322262.1 | NP_001309191.1 | 5 | 513 | |||
XM_011527440.2 | XP_011525742.1 | 6 | 446 | |||
GenBank mRNA | AK291549.1 | - | 6 | 560 | ||
BC035498.2 | - | 6 | 550 | 3841192 | ||
KF672847.1 | - | 4 | 389 | |||
KF672848.1 | - | 6 | 560 | |||
KR134287.1 | - | 6 | 560 | |||
M73812.1 | - | 4 | 389 | |||
M74093.1 | - | 4 | 357 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
