siRNA Details
Gene Symbol: FOS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005252.3 | NP_005243.1 | 2, 3 | 597 | ||
GenBank mRNA | AK290907.1 | - | 1, 2 | 485 | ||
AK291326.1 | - | 2, 3 | 547 | |||
BC004490.2 | - | 2, 3 | 518 | 3688670 | ||
BX647104.1 | - | 2, 3 | 547 | |||
CR541785.1 | - | 2, 3 | 392 | |||
CR542267.1 | - | 2, 3 | 392 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
