siRNA Details
Gene Symbol: STAT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003150.3 | NP_003141.2 | 5, 6 | 694 | ||
NM_139276.2 | NP_644805.1 | 5, 6 | 716 | |||
NM_213662.1 | NP_998827.1 | 5, 6 | 608 | |||
XM_005257616.3 | XP_005257673.2 | 5, 6 | 707 | |||
XM_005257617.3 | XP_005257674.2 | 5, 6 | 702 | |||
XM_011525145.2 | XP_011523447.1 | 5, 6 | 707 | |||
XM_011525146.2 | XP_011523448.1 | 5, 6 | 707 | |||
XM_017024972.1 | XP_016880461.1 | 5, 6 | 702 | |||
XM_017024973.1 | XP_016880462.1 | 5, 6 | 707 | |||
XM_017024974.1 | XP_016880463.1 | 5, 6 | 702 | |||
XM_017024975.1 | XP_016880464.1 | 5, 6 | 695 | |||
XM_017024976.1 | XP_016880465.1 | 5, 6 | 695 | |||
GenBank mRNA | AJ012463.1 | - | 4, 5 | 476 | ||
AK291933.1 | - | 5, 6 | 789 | |||
AK297994.1 | - | 4, 5 | 501 | |||
AK301200.1 | - | 5, 6 | 624 | |||
AK316139.1 | - | 5, 6 | 646 | |||
BC000627.2 | - | 5, 6 | 613 | 3347434 | ||
BC014482.1 | - | 5, 6 | 608 | 4909141 | ||
L29277.1 | - | 5, 6 | 696 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
