siRNA Details
Gene Symbol: BIRC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012271.1 | NP_001012271.1 | 4 | 423 | ||
NM_001168.2 | NP_001159.2 | 3 | 354 | |||
GenBank mRNA | AB028869.1 | - | 4 | 328 | ||
AB154416.1 | - | 3 | 233 | |||
AF077350.1 | - | 3 | 257 | |||
AK223428.1 | - | 3 | 299 | |||
AK301793.1 | - | 3 | 307 | |||
AK311917.1 | - | 3 | 258 | |||
AY830084.1 | - | 233 | ||||
BC008718.2 | - | 3 | 266 | 2961114 | ||
BC034148.1 | - | 3 | 277 | 4656567 | ||
BC065497.1 | - | 3 | 247 | 5394399 | ||
BI520186.1 | - | 4 | 433 | |||
BQ687404.1 | - | 3 | 131 | |||
CB112371.1 | - | 4 | 356 | |||
CR541740.1 | - | 3 | 233 | |||
DQ508245.1 | - | 4 | 238 | |||
DQ508246.1 | - | 4 | 234 | |||
DQ508248.1 | - | 4 | 266 | |||
DQ508249.1 | - | 3 | 883 | |||
HM625836.1 | - | 3 | 273 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
