siRNA Details
Gene Symbol: PLAUR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005376.2 | NP_001005376.1 | 4, 5 | 697 | ||
NM_001301037.1 | NP_001287966.1 | 4, 5 | 697 | |||
NM_002659.3 | NP_002650.1 | 4, 5 | 697 | |||
XM_005258990.4 | XP_005259047.1 | 4, 5 | 519 | |||
XM_011527027.2 | XP_011525329.1 | 3, 4 | 375 | |||
XM_011527028.2 | XP_011525330.1 | 4, 5 | 519 | |||
XM_011527029.1 | XP_011525331.1 | 4, 5 | 519 | |||
XM_011527031.2 | XP_011525333.1 | 4, 5 | 519 | |||
XM_017026872.1 | XP_016882361.1 | 4, 5 | 932 | |||
XM_017026873.1 | XP_016882362.1 | 2, 3 | 585 | |||
GenBank mRNA | A18757.1 | - | 4, 5 | 517 | ||
AF257789.1 | - | 4, 5 | 471 | |||
AK290774.1 | - | 4, 5 | 517 | |||
AY029180.1 | - | 4, 5 | 517 | |||
BC002788.2 | - | 4, 5 | 536 | 3617894 | ||
CA495321.1 | - | 4, 5 | 697 | |||
CF126152.1 | - | 4, 5 | 504 | |||
CR456952.1 | - | 4, 5 | 471 | |||
M83246.1 | - | 4, 5 | 488 | |||
X51675.1 | - | 4, 5 | 517 | |||
X74039.1 | - | 4, 5 | 517 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
