siRNA Details
Gene Symbol: SULT1A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177552.3 | NP_808220.1 | 8 | 1030 | ||
GenBank mRNA | BC078144.1 | - | 8 | 974 | 6022425 | |
BC132914.1 | - | 8 | 1027 | 40146936 | ||
BC132920.1 | - | 8 | 1027 | 40146942 | ||
BC144354.1 | - | 8 | 1027 | 9052874 | ||
BC144355.1 | - | 8 | 1027 | 9052875 | ||
BC171772.1 | - | 8 | 1027 | 9054426 | ||
L25275.1 | - | 8 | 990 | |||
U08032.1 | - | 7 | 932 |
Gene Symbol: SLX1A-SULT1A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037608.1 | 11 | 1847 | |||
GenBank mRNA | AK094769.1 | - | 9 | 2329 | ||
AK097386.1 | - | 8 | 1490 | |||
AK122707.1 | - | 11 | 1848 | |||
AK122733.1 | - | 10 | 2266 | |||
BC014471.1 | - | 9 | 1112 | 4907290 | ||
BC111011.1 | - | 10 | 1875 | 5768190 |
Gene Symbol: SLX1B-SULT1A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037609.1 | 11 | 1847 |
Gene Symbol: SULT1A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017390.2 | NP_001017390.1 | 8 | 1030 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
