siRNA Details
Gene Symbol: TUB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003320.4 | NP_003311.2 | 1 | 348 | ||
GenBank mRNA | BC075031.2 | - | 1 | 140 | 30915625 | |
BC075032.2 | - | 1 | 140 | 30915418 | ||
U82467.1 | - | 1 | 348 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
