siRNA Details
Gene Symbol: EPB41L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258329.1 | NP_001245258.1 | 3 | 354 | ||
NM_012156.2 | NP_036288.2 | 2 | 300 | |||
XM_011528671.1 | XP_011526973.1 | 3 | 333 | |||
XM_011528673.1 | XP_011526975.1 | 2 | 293 | |||
XM_011528681.1 | XP_011526983.1 | 1 | 143 | |||
XM_011528682.1 | XP_011526984.1 | 1 | 143 | |||
XM_011528683.1 | XP_011526985.1 | 1 | 143 | |||
XM_011528685.1 | XP_011526987.1 | 1 | 143 | |||
XM_017027715.1 | XP_016883204.1 | 1 | 143 | |||
XM_017027716.1 | XP_016883205.1 | 3 | 333 | |||
XM_017027717.1 | XP_016883206.1 | 2 | 291 | |||
XM_017027720.1 | XP_016883209.1 | 4 | 404 | |||
GenBank mRNA | AB002336.1 | - | 2 | 290 | ||
AK299817.1 | - | 2 | 247 | |||
AY049789.1 | - | 1 | 138 | |||
BC040259.1 | - | 3 | 332 | 5195986 | ||
CR936825.1 | - | 3 | 357 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
