siRNA Details
Gene Symbol: KAT14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020536.4 | NP_065397.3 | 1 | 521 | ||
GenBank mRNA | BC007537.1 | - | 1 | 425 | 3350378 | |
BM552422.1 | - | 2 | 719 | |||
BQ691686.1 | - | 1 | 521 |
Gene Symbol: PET117 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164811.1 | NP_001158283.1 | 2 | 734 | ||
GenBank mRNA | AY358666.1 | - | 2 | 641 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
