siRNA Details
Gene Symbol: HMCN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291815.1 | NP_001278744.1 | 96, 97 | 14765 | ||
XM_011518465.2 | XP_011516767.1 | 95, 96 | 14779 | |||
XM_011518466.2 | XP_011516768.1 | 95, 96 | 14770 | |||
XM_011518467.2 | XP_011516769.1 | 95, 96 | 14725 | |||
XM_017014585.1 | XP_016870074.1 | 75, 76 | 11571 | |||
XM_017014586.1 | XP_016870075.1 | 50, 51 | 7499 | |||
GenBank mRNA | AK074396.1 | - | 2, 3 | 442 | ||
AK128680.1 | - | 1, 2 | 2319 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
