siRNA Details
Gene Symbol: NAPRT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286829.1 | NP_001273758.1 | 11 | 1358 | ||
NM_145201.5 | NP_660202.3 | 11 | 1358 | |||
XM_017013975.1 | XP_016869464.1 | 10 | 2920 | |||
XM_017013976.1 | XP_016869465.1 | 10 | 2914 | |||
XM_017013977.1 | XP_016869466.1 | 10 | 2620 | |||
XM_017013979.1 | XP_016869468.1 | 10 | 1116 | |||
XM_017013980.1 | XP_016869469.1 | 10 | 1221 | |||
GenBank mRNA | AB242230.1 | - | 11 | 1345 | ||
AF258565.1 | - | 5 | 806 | |||
AK097002.1 | - | 11 | 1350 | |||
AY214325.1 | - | 11 | 1346 | |||
AY214326.1 | - | 11 | 1346 | |||
AY214327.1 | - | 11 | 1346 | |||
BC006284.2 | - | 11 | 1330 | 3957135 | ||
BC014190.2 | - | 11 | 1330 | |||
BC032466.1 | - | 6 | 644 | 5210589 |
