siRNA Details
Gene Symbol: VPS13B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015243.2 | NP_056058.2 | 15 | 2229 | ||
NM_017890.4 | NP_060360.3 | 15 | 2229 | |||
NM_152564.4 | NP_689777.3 | 15 | 2229 | |||
XM_005250800.3 | XP_005250857.1 | 15 | 2535 | |||
XM_005250801.4 | XP_005250858.1 | 15 | 8405 | |||
XM_006716510.3 | XP_006716573.1 | 15 | 2221 | |||
XM_011516848.2 | XP_011515150.1 | 15 | 2221 | |||
XM_011516849.2 | XP_011515151.1 | 15 | 2221 | |||
XM_011516850.2 | XP_011515152.1 | 13 | 1861 | |||
XM_011516853.2 | XP_011515155.1 | 15 | 2221 | |||
XM_011516859.2 | XP_011515161.1 | 15 | 2221 | |||
XM_011516861.1 | XP_011515163.1 | 15 | 2221 | |||
XM_011516865.1 | XP_011515167.1 | 15 | 2221 | |||
XM_011516866.2 | XP_011515168.1 | 15 | 2221 | |||
XM_017013110.1 | XP_016868599.1 | 8 | 1019 | |||
GenBank mRNA | AJ608772.1 | - | 15 | 2221 | ||
AJ608773.1 | - | 15 | 2221 | |||
AK091431.1 | - | 4 | 569 | |||
AY223814.1 | - | 15 | 2229 | |||
AY223815.1 | - | 15 | 2229 | |||
AY223816.1 | - | 15 | 2229 | |||
AY223817.1 | - | 15 | 2229 | |||
BC033121.1 | - | 1 | 113 | |||
BX647561.1 | - | 1 | 54 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
