siRNA Details
Gene Symbol: LINC00205 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF426264.1 | - | 2 | 698 |
Gene Symbol: LOC107985476 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017028526.1 | XP_016884015.1 | 1 | 165 |
