siRNA Details
Gene Symbol: ANKRD30BP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026916.1 | 6 | 811 | |||
GenBank mRNA | AF427490.1 | - | 4 | 432 | ||
BC126176.1 | - | 3 | 270 | |||
BC126178.1 | - | 3 | 270 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
