siRNA Details
Gene Symbol: RRP7BP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002184.2 | 6 | 720 | |||
GenBank mRNA | AK074492.1 | - | 4 | 481 | ||
AK098201.1 | - | 6 | 747 | |||
AL157851.1 | - | 6 | 705 | |||
BC007736.2 | - | 6 | 720 | |||
BC014647.1 | - | 6 | 718 | |||
BC143340.1 | - | 6 | 808 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
