siRNA Details
Gene Symbol: NTRK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012331.1 | NP_001012331.1 | 1, 2 | 262 | ||
NM_002529.3 | NP_002520.2 | 1, 2 | 262 | |||
GenBank mRNA | AK290759.1 | - | 1, 2 | 247 | ||
AK312704.1 | - | 1, 2 | 247 | |||
BC062580.1 | - | 1, 2 | 205 | |||
BC136554.1 | - | 1, 2 | 306 | 9020544 | ||
BC144239.1 | - | 1, 2 | 306 | 9052759 | ||
KJ534902.1 | - | 1, 2 | 206 | |||
M23102.1 | - | 1, 2 | 326 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
