siRNA Details
Gene Symbol: ABCA13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152701.4 | NP_689914.3 | 62 | 16428 | ||
XM_011515130.2 | XP_011513432.1 | 62 | 16463 | |||
XM_011515131.2 | XP_011513433.1 | 62 | 16370 | |||
XM_011515132.2 | XP_011513434.1 | 61 | 16370 | |||
XM_011515133.2 | XP_011513435.1 | 61 | 16325 | |||
XM_011515136.2 | XP_011513438.2 | 57 | 15828 | |||
GenBank mRNA | AK128516.1 | - | 19 | 3723 | ||
AY204751.1 | - | 62 | 16429 |
Gene Symbol: CASC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK098265.1 | - | 2 | 814 |
Gene Symbol: DAND5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152654.2 | NP_689867.1 | 2 | 1061 | ||
GenBank mRNA | AK095926.1 | - | 2 | 1060 | ||
BC025333.1 | - | 4 | 1287 |
Gene Symbol: DNAJA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK127660.1 | - | 1 | 455 |
Gene Symbol: LOC440896 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015361.1 | 3 | 1779 | |||
GenBank mRNA | AK055091.1 | - | 3 | 1782 | ||
AK127288.1 | - | 3 | 2893 | |||
BC036611.1 | - | 2 | 1399 |
Gene Symbol: PEX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000318.2 | NP_000309.1 | 4 | 2028 | ||
NM_001079867.1 | NP_001073336.1 | 3 | 1996 | |||
NM_001172086.1 | NP_001165557.1 | 5 | 2034 | |||
NM_001172087.1 | NP_001165558.1 | 3 | 1892 |
Gene Symbol: SEH1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011525746.2 | XP_011524048.1 | 1 | 121 | ||
XM_017026026.1 | XP_016881515.1 | 1 | 82 | |||
XM_017026027.1 | XP_016881516.1 | 1 | 185 |
Gene Symbol: SNHG16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038108.1 | 4 | 1170 | |||
NR_038109.1 | 4 | 1166 | ||||
NR_038110.1 | 3 | 1071 | ||||
NR_038111.1 | 3 | 1067 | ||||
GenBank mRNA | AB073602.1 | - | 4 | 1030 | ||
AK024090.1 | - | 2 | 510 | |||
AK055935.1 | - | 5 | 1098 | |||
AK129904.1 | - | 4 | 1036 | |||
BC042949.2 | - | 3 | 2239 | |||
BC112893.1 | - | 2 | 843 | |||
BX647993.1 | - | 3 | 1026 |
Gene Symbol: TRIM59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173084.2 | NP_775107.1 | 3 | 2783 |
Gene Symbol: ZNF493 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145326.2 | NP_663299.2 | 4 | 412 | ||
GenBank mRNA | BC006408.1 | - | 4 | 375 |
Gene Symbol: ZNF595 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK309855.1 | - | 4 | 633 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
