siRNA Details
Gene Symbol: MAFK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002360.3 | NP_002351.1 | 3 | 1848 | ||
XM_006715773.2 | XP_006715836.1 | 3 | 1901 | |||
GenBank mRNA | AK056767.1 | - | 1 | 717 | ||
AL831913.1 | - | 1 | 565 | |||
BC012777.2 | - | 1 | 105 | |||
BC143040.1 | - | 1 | 1140 | |||
BC143041.1 | - | 1 | 1142 | |||
BC148194.1 | - | 1 | 1142 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
