siRNA Details
Gene Symbol: H2AFJ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177925.3 | NP_808760.1 | 1 | 1276 | ||
NR_027716.2 | 2 | 962 | ||||
GenBank mRNA | AK001765.1 | - | 2 | 722 | ||
AK303301.1 | - | 3 | 839 | |||
AL133626.1 | - | 2 | 716 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
