siRNA Details
Gene Symbol: DGKZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105540.1 | NP_001099010.1 | 4, 5 | 1059 | ||
NM_001199267.1 | NP_001186196.1 | 3, 4 | 621 | |||
NM_201532.2 | NP_963290.1 | 3, 4 | 789 | |||
XM_006718355.2 | XP_006718418.1 | 3, 4 | 397 | |||
XM_017018456.1 | XP_016873945.1 | 4, 5 | 1217 | |||
GenBank mRNA | AK124594.1 | - | 3, 4 | 789 | ||
AK126970.1 | - | 3, 4 | 505 | |||
BX648433.1 | - | 3, 4 | 356 | |||
U51477.1 | - | 3, 4 | 455 | |||
U94905.1 | - | 4, 5 | 1059 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
