siRNA Details
Gene Symbol: MAFK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002360.3 | NP_002351.1 | 3 | 3001 | ||
XM_006715773.2 | XP_006715836.1 | 3 | 3054 | |||
GenBank mRNA | AK056767.1 | - | 1 | 1870 | ||
AL831913.1 | - | 1 | 1718 | |||
BC012777.2 | - | 1 | 1258 | |||
BC094871.1 | - | 1 | 917 |
Gene Symbol: TMEM184A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BM981655.1 | - | 1 | 476 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
