siRNA Details
Gene Symbol: RPSA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304288.1 | NP_001291217.1 | 4 | 540 | ||
NM_002295.5 | NP_002286.2 | 4 | 525 | |||
GenBank mRNA | AF503367.1 | - | 1 | 485 | ||
AK055991.1 | - | 1 | 659 | |||
BC005391.1 | - | 1 | 497 | 3997019 | ||
BC008867.2 | - | 1 | 446 | 4130936 | ||
BC010418.2 | - | 1 | 477 | 4079845 | ||
BC013827.2 | - | 1 | 481 | 3446816 | ||
BC018867.2 | - | 1 | 471 | |||
BC034537.1 | - | 1 | 478 | 3455545 | ||
BC050688.1 | - | 1 | 480 | 4443949 | ||
BC053370.1 | - | 1 | 478 | 5108337 | ||
BC062714.1 | - | 1 | 501 | 3686924 | ||
BC066941.1 | - | 4 | 494 | 4820432 | ||
BC068062.1 | - | 1 | 459 | 5427114 | ||
BC070263.1 | - | 4 | 525 | 3996864 | ||
BC071693.1 | - | 1 | 487 | 3884457 | ||
BC071968.1 | - | 1 | 478 | 5814758 | ||
BC071969.1 | - | 1 | 466 | 6483856 | ||
BC071970.1 | - | 1 | 451 | 6484091 | ||
BC073863.1 | - | 1 | 463 | 3924066 | ||
BC107567.1 | - | 1 | 498 | 3681016 | ||
BT007219.1 | - | 3 | 416 | |||
J03799.1 | - | 3 | 453 | |||
S37431.1 | - | 3 | 431 | |||
X15005.1 | - | 4 | 494 | |||
X61156.1 | - | 3 | 386 |
Gene Symbol: RPSAP15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF284768.1 | - | 1 | 493 |
Gene Symbol: RPSAP19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX648992.1 | - | 2 | 1971 |
Gene Symbol: RPSAP58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003662.3 | 4 | 839 |
Gene Symbol: RPSAP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026890.1 | 1 | 516 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
