siRNA Details
Gene Symbol: DAND5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152654.2 | NP_689867.1 | 2 | 1292 | ||
GenBank mRNA | AK095926.1 | - | 2 | 1291 | ||
BC025333.1 | - | 4 | 1516 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
