siRNA Details
Gene Symbol: NOMO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004060.1 | NP_001004060.1 | 13, 14 | 1605 | ||
NM_173614.2 | NP_775885.1 | 13, 14 | 1605 | |||
XM_017023156.1 | XP_016878645.1 | 13, 14 | 1652 | |||
GenBank mRNA | AK225803.1 | - | 13, 14 | 1575 | ||
AL832855.1 | - | 13, 14 | 1599 | |||
BC028389.1 | - | 12, 13 | 1465 | 4824724 | ||
BC041131.1 | - | 13, 14 | 1579 | 5757234 |
Gene Symbol: NOMO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014287.3 | NP_055102.3 | 13, 14 | 1599 | ||
GenBank mRNA | BC065535.1 | - | 13, 14 | 1552 | 6007603 |
Gene Symbol: NOMO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004067.3 | NP_001004067.1 | 13, 14 | 1668 | ||
XM_005255318.1 | XP_005255375.1 | 13, 14 | 1662 | |||
GenBank mRNA | BC006531.2 | - | 2, 3 | 289 | ||
BM507095.1 | - | 4, 5 | 536 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
