siRNA Details
Gene Symbol: COX18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297732.1 | NP_001284661.1 | 6 | 1912 | ||
NM_001297733.1 | NP_001284662.1 | 6 | 1913 | |||
NM_001300729.1 | NP_001287658.1 | 5 | 1808 | |||
NM_173827.3 | NP_776188.1 | 6 | 1909 | |||
XM_005265680.4 | XP_005265737.1 | 6 | 1916 | |||
XM_011531878.2 | XP_011530180.1 | 6 | 1883 | |||
XM_017008045.1 | XP_016863534.1 | 6 | 1913 | |||
GenBank mRNA | AK096310.2 | - | 6 | 1855 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
