siRNA Details
Gene Symbol: PAPPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002581.4 | NP_002572.2 | 22 | 9504 | ||
XM_006717129.2 | XP_006717192.1 | 20 | 7578 | |||
XM_017014784.1 | XP_016870273.1 | 21 | 9430 | |||
GenBank mRNA | AY140907.1 | - | 1 | 787 | ||
AY189937.1 | - | 1 | 79 | |||
AY623010.1 | - | 1 | 438 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
