siRNA Details
Gene Symbol: MMP19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001272101.1 | NP_001259030.1 | 5 | 703 | ||
NM_002429.5 | NP_002420.1 | 6 | 949 | |||
NR_073606.1 | 6 | 920 | ||||
XM_006719401.3 | XP_006719464.1 | 4 | 590 | |||
XM_011538359.1 | XP_011536661.1 | 5 | 689 | |||
XM_017019308.1 | XP_016874797.1 | 6 | 935 | |||
XM_017019309.1 | XP_016874798.1 | 6 | 935 | |||
GenBank mRNA | AK297999.1 | - | 4 | 529 | ||
AK303202.1 | - | 5 | 656 | |||
BC050368.1 | - | 6 | 888 | 5761074 | ||
BI761698.1 | - | 4 | 540 | |||
U37791.1 | - | 6 | 877 | |||
U38320.1 | - | 2 | 566 | |||
U38321.1 | - | 6 | 872 | |||
U38322.1 | - | 2 | 577 | |||
U38431.1 | - | 6 | 841 | |||
X92521.1 | - | 6 | 897 | |||
Y08622.1 | - | 6 | 896 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
