siRNA Details
Gene Symbol: RAB37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175738.4 | NP_783865.1 | 2 | 601 | ||
GenBank mRNA | AK054846.1 | - | 2 | 538 | ||
AK127073.1 | - | 2 | 1140 | |||
AK290202.1 | - | 2 | 577 | |||
BC040547.1 | - | 2 | 587 | 5287038 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
