siRNA Details
Gene Symbol: CXorf36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176819.3 | NP_789789.2 | 4, 5 | 1037 | ||
XM_005272670.1 | XP_005272727.1 | 3, 4 | 838 | |||
GenBank mRNA | AK094877.1 | - | 2, 3 | 262 | ||
AK294472.1 | - | 3, 4 | 606 | |||
DA378381.1 | - | 1, 2 | 131 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
