siRNA Details
Gene Symbol: TAS2R19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176888.2 | NP_795369.1 | 1 | 177 | ||
GenBank mRNA | BC101804.1 | - | 1 | 177 | 8069310 | |
BC111998.1 | - | 1 | 189 | 8327466 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
