siRNA Details
Gene Symbol: TAS2R50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176890.2 | NP_795371.2 | 1 | 171 | ||
GenBank mRNA | BC112100.1 | - | 1 | 171 | 8327568 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
