siRNA Details
Gene Symbol: ATL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290048.1 | NP_001276977.1 | 13 | 5374 | ||
NM_015459.4 | NP_056274.3 | 13 | 5476 | |||
XM_006718493.1 | XP_006718556.1 | 12 | 5301 | |||
GenBank mRNA | AK090822.1 | - | 1 | 561 | ||
BX648468.1 | - | 1 | 1989 | |||
CR936784.1 | - | 13 | 5346 |
Gene Symbol: CFAP157 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK094948.1 | - | 8 | 1995 |
Gene Symbol: SMCHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015295.2 | NP_056110.2 | 48 | 8061 | ||
XM_011525642.1 | XP_011523944.1 | 49 | 8164 | |||
XM_011525644.2 | XP_011523946.1 | 48 | 8072 | |||
XM_017025684.1 | XP_016881173.1 | 50 | 8342 | |||
GenBank mRNA | AB014550.1 | - | 22 | 4399 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
