siRNA Details
Gene Symbol: CXorf36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176819.3 | NP_789789.2 | 5 | 1172 | ||
XM_005272670.1 | XP_005272727.1 | 4 | 973 | |||
XM_006724559.1 | XP_006724622.1 | 4 | 895 | |||
GenBank mRNA | AK094877.1 | - | 3 | 397 | ||
AK294472.1 | - | 4 | 741 | |||
DA378381.1 | - | 2 | 266 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
