siRNA Details
Gene Symbol: TAS2R30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001097643.1 | NP_001091112.1 | 1 | 228 |
Gene Symbol: TAS2R31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176885.2 | NP_795366.2 | 1 | 300 | ||
GenBank mRNA | BC117421.1 | - | 1 | 301 | 40125972 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
