siRNA Details
Gene Symbol: NOMO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004060.1 | NP_001004060.1 | 32 | 3762 | ||
NM_173614.2 | NP_775885.1 | 31 | 4093 | |||
GenBank mRNA | AL832855.1 | - | 32 | 3756 | ||
BC028389.1 | - | 31 | 3622 | 4824724 | ||
BC041131.1 | - | 31 | 4067 | 5757234 |
Gene Symbol: LOC102723728 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006720996.3 | XP_006721059.1 | 9 | 1401 |
Gene Symbol: NOMO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004067.3 | NP_001004067.1 | 31 | 4156 | ||
XM_005255318.1 | XP_005255375.1 | 32 | 3819 | |||
GenBank mRNA | BC006531.2 | - | 20 | 2777 | ||
BC013837.1 | - | 5 | 917 | |||
BQ948282.1 | - | 2 | 516 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
