siRNA Details
Gene Symbol: VN1R3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF336873.1 | - | 1 | 247 | ||
BC107073.1 | - | 1 | 247 | |||
BC107074.1 | - | 1 | 247 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
