siRNA Details
Gene Symbol: ATL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290048.1 | NP_001276977.1 | 13 | 5298 | ||
NM_015459.4 | NP_056274.3 | 13 | 5400 | |||
XM_006718493.1 | XP_006718556.1 | 12 | 5225 | |||
GenBank mRNA | AK090822.1 | - | 1 | 485 | ||
BX648468.1 | - | 1 | 1913 | |||
CR936784.1 | - | 13 | 5270 |
Gene Symbol: AOAH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011515339.2 | XP_011513641.1 | 20 | 5586 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
