siRNA Details
Gene Symbol: PDLIM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021630.5 | NP_067643.3 | 9 | 1643 | ||
NM_176871.3 | NP_789847.1 | 8 | 985 | |||
GenBank mRNA | AF289563.1 | - | 9 | 1243 | ||
AK024498.1 | - | 8 | 928 | |||
AK027800.1 | - | 9 | 1254 | |||
AK056748.1 | - | 3 | 1410 | |||
AK074031.1 | - | 2 | 2695 | |||
AK090432.1 | - | 1 | 3910 | |||
AK092968.1 | - | 9 | 1643 | |||
AY007729.1 | - | 7 | 928 | |||
AY217349.1 | - | 8 | 970 | |||
BC021556.1 | - | 9 | 1124 | 3959372 | ||
BC071774.1 | - | 9 | 1254 | 4593207 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
