siRNA Details
Gene Symbol: TAS2R43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176884.2 | NP_795365.2 | 1 | 203 | ||
GenBank mRNA | BC101155.1 | - | 2 | 119 | 40015679 | |
BC117423.1 | - | 1 | 202 | 40125974 |
Gene Symbol: TAS2R30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001097643.1 | NP_001091112.1 | 1 | 119 |
Gene Symbol: TAS2R45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176886.2 | NP_795367.2 | 2 | 119 | ||
GenBank mRNA | BC101154.1 | - | 2 | 119 | 40015678 | |
BC101156.1 | - | 2 | 119 | 40015680 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
