siRNA Details
Gene Symbol: TAS2R43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176884.2 | NP_795365.2 | 1 | 888 | ||
GenBank mRNA | BC117423.1 | - | 1 | 887 | 40125974 |
Gene Symbol: TAS2R31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176885.2 | NP_795366.2 | 1 | 876 | ||
GenBank mRNA | BC117421.1 | - | 1 | 877 | 40125972 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
