siRNA Details
Gene Symbol: MAFB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005461.4 | NP_005452.2 | 1 | 482 | ||
GenBank mRNA | AF134157.1 | - | 1 | 165 | ||
AK027324.1 | - | 1 | 100 | |||
BC028098.1 | - | 1 | 113 | 5242606 | ||
BC036689.1 | - | 1 | 483 | 5261053 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
