siRNA Details
Gene Symbol: ZNF271P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024565.1 | 3 | 1068 | |||
NR_024566.1 | 2 | 845 | ||||
GenBank mRNA | AF159567.1 | - | 2 | 785 | ||
AK023456.1 | - | 3 | 1011 | |||
AK094216.1 | - | 2 | 1578 | |||
BC017710.1 | - | 1 | 263 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
