siRNA Details
Gene Symbol: ST8SIA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015879.2 | NP_056963.2 | 1 | 451 | ||
GenBank mRNA | AB208996.1 | - | 1 | 455 | ||
AF003092.2 | - | 1 | 166 | |||
AF004668.2 | - | 1 | 2117 | |||
AK289517.1 | - | 1 | 229 | |||
AK313047.1 | - | 1 | 177 | |||
BC074909.2 | - | 1 | 142 | 30915256 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
