siRNA Details
Gene Symbol: PPP1CB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002709.2 | NP_002700.1 | 2 | 479 | ||
NM_206876.1 | NP_996759.1 | 3 | 274 | |||
GenBank mRNA | AF092905.1 | - | 4 | 252 | ||
AK295957.1 | - | 2 | 137 | |||
AK303682.1 | - | 2 | 138 | |||
AK312329.1 | - | 2 | 416 | |||
AK316532.1 | - | 2 | 100 | |||
BC002697.2 | - | 2 | 195 | 3609386 | ||
BC012045.1 | - | 2 | 195 | 3631338 | ||
BI551898.1 | - | 3 | 281 | |||
BT019744.1 | - | 2 | 114 | |||
BX477003.1 | - | 2 | 423 | |||
BX647970.1 | - | 3 | 313 | |||
CD656807.1 | - | 3 | 241 | |||
CR542263.1 | - | 2 | 114 | |||
CR542285.1 | - | 2 | 114 | |||
EU668339.1 | - | 2 | 195 | |||
X80910.1 | - | 2 | 372 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
