siRNA Details
Gene Symbol: NLGN4Y | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206850.1 | NP_001193779.1 | 5 | 1446 | ||
NM_014893.4 | NP_055708.3 | 5 | 1776 | |||
NR_028319.1 | 5 | 1998 | ||||
NR_046355.1 | 6 | 1763 | ||||
XM_006724874.2 | XP_006724937.1 | 6 | 2008 | |||
XM_011531424.2 | XP_011529726.1 | 6 | 2326 | |||
XM_011531425.2 | XP_011529727.1 | 6 | 2098 | |||
XM_011531426.2 | XP_011529728.1 | 6 | 2059 | |||
XM_011531427.1 | XP_011529729.1 | 6 | 1813 | |||
XM_011531428.1 | XP_011529730.1 | 7 | 1994 | |||
XM_011531429.2 | XP_011529731.1 | 6 | 2609 | |||
XM_011531430.2 | XP_011529732.1 | 6 | 1947 | |||
XM_017030034.1 | XP_016885523.1 | 5 | 2038 | |||
XM_017030035.1 | XP_016885524.1 | 5 | 1959 | |||
XM_017030036.1 | XP_016885525.1 | 5 | 1753 | |||
XM_017030037.1 | XP_016885526.1 | 5 | 1751 | |||
XM_017030038.1 | XP_016885527.1 | 5 | 2549 | |||
XM_017030039.1 | XP_016885528.1 | 6 | 1284 | |||
XM_017030040.1 | XP_016885529.1 | 5 | 1133 | |||
GenBank mRNA | AB023168.1 | - | 5 | 1302 | ||
AF376804.1 | - | 5 | 1776 | |||
AK295117.1 | - | 6 | 1763 | |||
BC113525.1 | - | 4 | 1603 | 8322577 | ||
BC113551.1 | - | 4 | 1603 | 8322603 | ||
BX537428.1 | - | 5 | 1999 |
