siRNA Details
Gene Symbol: ATAD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014109.3 | NP_054828.2 | 27 | 4169 | ||
XM_011516994.2 | XP_011515296.1 | 26 | 4306 | |||
XM_011516996.2 | XP_011515298.1 | 16 | 2589 | |||
GenBank mRNA | AF118088.1 | - | 10 | 1627 | ||
AL833653.1 | - | 27 | 3923 | |||
AY598335.1 | - | 27 | 4061 | |||
BC010686.1 | - | 12 | 2037 | |||
BC019909.1 | - | 9 | 1373 | |||
BC113656.1 | - | 27 | 4083 | 8322708 | ||
CR749832.1 | - | 28 | 4398 |
