siRNA Details
Gene Symbol: UBE2J2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_058167.2 | NP_477515.2 | 3, 4 | 369 | ||
NM_194315.1 | NP_919296.1 | 4, 5 | 417 | |||
NM_194457.1 | NP_919439.1 | 2, 3 | 238 | |||
NM_194458.1 | NP_919440.1 | 3, 4 | 498 | |||
XM_005244718.3 | XP_005244775.1 | 3, 4 | 647 | |||
XM_005244719.4 | XP_005244776.1 | 3, 4 | 535 | |||
XM_006710333.3 | XP_006710396.1 | 3, 4 | 814 | |||
XM_011540613.2 | XP_011538915.1 | 2, 3 | 242 | |||
XM_011540614.2 | XP_011538916.1 | 3, 4 | 600 | |||
XM_017000239.1 | XP_016855728.1 | 2, 3 | 698 | |||
GenBank mRNA | AF296658.1 | - | 3, 4 | 322 | ||
AK056065.1 | - | 3, 4 | 279 | |||
AK075017.1 | - | 3, 4 | 348 | |||
AK291935.1 | - | 3, 4 | 348 | |||
BC094777.1 | - | 3, 4 | 368 | 30340049 | ||
BC104890.1 | - | 2, 3 | 193 | 8143893 | ||
BC113430.1 | - | 2, 3 | 193 | 8322482 | ||
BC143657.1 | - | 2, 3 | 193 | 9052171 | ||
BE382927.1 | - | 3, 4 | 312 | |||
BG112933.1 | - | 493 | ||||
BG499172.1 | - | 3, 4 | 362 | |||
BM544827.1 | - | 4, 5 | 420 | |||
BM765278.1 | - | 2, 3 | 153 | |||
BM926567.1 | - | 3, 4 | 495 | |||
BQ881730.1 | - | 1, 2 | 58 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
